accessibility menu, dialog, popup

UserName

NH2BrFFF

CAS RN 57946-63-1

IUPAC Name: 2-bromo-4-(trifluoromethyl)aniline
Synonyms: 2-bromo-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.02
Molecular Formula: C7H5BrF3N
InChi Key: QKRJIXSZTKOFTD-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Br)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results