missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%

Catalog No. p-7024752
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1994906 5 g
AAA1994914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1994906 Supplier Thermo Scientific Chemicals Supplier No. A1994906

CAS: 770-30-9 | C6H6N4S | 166.20 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 770-30-9
Molecular Formula C6H6N4S
Molecular Weight (g/mol) 166.20
MDL Number MFCD00023237
InChI Key VYDHBDPZNLVELI-UHFFFAOYSA-N
Synonym 4-amino-2-methylthio pyrimidine-5-carbonitrile, 4-amino-2-methylsulfanyl pyrimidine-5-carbonitrile, 4-amino-2-methylthio-5-pyrimidinecarbonitrile, 4-amino-2-methylsulfanyl-pyrimidine-5-carbonitrile, 4-amino-2-methylthiopyrimidine-5-carbonitrile, maybridge1_004789, acmc-20a4j5, 4-amino-5-cyano-2-methylthiopyrimidine, 4-amino-5-cyano-2-methylthio pyrimidine
PubChem CID 295757
IUPAC Name 4-amino-2-methylsulfanylpyrimidine-5-carbonitrile
SMILES CSC1=NC=C(C#N)C(N)=N1

Specifications

Melting Point 240°C to 243°C
Quantity 5 g
UN Number UN3439
Solubility Information Soluble in water.
Formula Weight 166.21
Percent Purity 97%
Chemical Name or Material 4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only