missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-methylbenzoic Acid 98.0+%, TCI America™
$55.92 - $223.68
Chemical Identifiers
| CAS | 2486-75-1 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD06656133 |
| InChI Key | XRSQZFJLEPBPOZ-UHFFFAOYSA-N |
| Synonym | 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid |
| PubChem CID | 241632 |
| IUPAC Name | 4-amino-2-methylbenzoic acid |
| SMILES | CC1=C(C=CC(=C1)N)C(=O)O |
Chemical Identifiers
| 2486-75-1 | |
| 151.165 | |
| XRSQZFJLEPBPOZ-UHFFFAOYSA-N | |
| 241632 | |
| CC1=C(C=CC(=C1)N)C(=O)O |
| C8H9NO2 | |
| MFCD06656133 | |
| 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid | |
| 4-amino-2-methylbenzoic acid |
Specifications
| 2486-75-1 | |
| Yellow | |
| MFCD06656133 | |
| 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid | |
| CC1=C(C=CC(=C1)N)C(=O)O | |
| 151.165 | |
| 151.17 | |
| Crystalline Powder |
| 164°C | |
| C8H9NO2 | |
| 5 g | |
| XRSQZFJLEPBPOZ-UHFFFAOYSA-N | |
| 4-amino-2-methylbenzoic acid | |
| 241632 | |
| ≥98.0% (HPLC,T) | |
| 4-Amino-2-methylbenzoic Acid |
Safety and Handling
TSCA : No