missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-methylbenzoic Acid 98.0+%, TCI America™
Supplier: TCI America A234925G
Specifications
| 4-Amino-2-methylbenzoic Acid | |
| 164°C | |
| C8H9NO2 | |
| 25 g | |
| XRSQZFJLEPBPOZ-UHFFFAOYSA-N | |
| 4-amino-2-methylbenzoic acid | |
| 241632 | |
| ≥98.0% (HPLC,T) |
| 2486-75-1 | |
| Yellow | |
| MFCD06656133 | |
| 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid | |
| CC1=C(C=CC(=C1)N)C(=O)O | |
| 151.165 | |
| 151.17 | |
| Crystalline Powder |
Chemical Identifiers
| 2486-75-1 | |
| 151.165 | |
| XRSQZFJLEPBPOZ-UHFFFAOYSA-N | |
| 241632 | |
| CC1=C(C=CC(=C1)N)C(=O)O |
| C8H9NO2 | |
| MFCD06656133 | |
| 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid | |
| 4-amino-2-methylbenzoic acid |
Safety and Handling
TSCA : No