missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methoxyphenol 98.0+%, TCI America™

Catalog No. p-7118125
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A28831G 1 g
A28835G 5 g
2 options

Catalog No. A28831G

Supplier: TCI America A28831G

Only null left

Chemical Identifiers

CAS 52200-90-5
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD01707809
InChI Key MCNBYOWWTITHIG-UHFFFAOYSA-N
Synonym 4-Hydroxy-3-methoxyaniline, 4-Aminoguaiacol
PubChem CID 101296
IUPAC Name 4-amino-2-methoxyphenol
SMILES COC1=C(C=CC(=C1)N)O

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 139.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methoxyphenol
RTECSNumber SJ6072000
TSCA No