CAS RN 52200-90-5
IUPAC Name:
4-amino-2-methoxyphenol
Synonyms:
4-amino-2-methoxyphenol
Molecular Weight (g/mol):
139.15
Molecular Formula:
C7H9NO2
InChi Key:
MCNBYOWWTITHIG-UHFFFAOYSA-N
SMILES:
COC1=CC(N)=CC=C1O
1
–
1
of
1
results
1
4-Amino-2-methoxyphenol 98.0+%, TCI America™
CAS: 52200-90-5 Molecular Formula: C7H9NO2 Molecular Weight (g/mol): 139.154 MDL Number: MFCD01707809 InChI Key: MCNBYOWWTITHIG-UHFFFAOYSA-N Synonym: 4-Hydroxy-3-methoxyaniline, 4-Aminoguaiacol PubChem CID: 101296 IUPAC Name: 4-amino-2-methoxyphenol SMILES: COC1=C(C=CC(=C1)N)O