missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methoxyphenol 98.0+%, TCI America™

Catalog No. A28835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28835G 5 g
A28831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28835G Supplier TCI America Supplier No. A28835G
Only null left

Chemical Identifiers

CAS 52200-90-5
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD01707809
InChI Key MCNBYOWWTITHIG-UHFFFAOYSA-N
Synonym 4-Hydroxy-3-methoxyaniline, 4-Aminoguaiacol
PubChem CID 101296
IUPAC Name 4-amino-2-methoxyphenol
SMILES COC1=C(C=CC(=C1)N)O

Specifications

Melting Point 180°C
Quantity 5 g
Formula Weight 139.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methoxyphenol
RTECSNumber SJ6072000
TSCA No