missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. A284025G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A284025G 25 g
A28405G 5 g
2 options

Catalog No. A284025G

Supplier: TCI America A284025G

Only null left

Chemical Identifiers

CAS 2486-80-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00114479
InChI Key OLJXRTRRJSMURJ-UHFFFAOYSA-N
Synonym 4-amino-o-anisic acid, 4-amino-2-methoxy-benzoic acid, 4-amino-2-methoxybenzenecarboxylic acid, 2-methoxy-4-amino-benzoic acid, 4-amino-2-methoxybenzoicacid, 4-azanyl-2-methoxy-benzoic acid, benzoic acid, 4-amino-2-methoxy, 2-methoxy-4-aminobenzoic acid, acmc-20aicb, pubchem4934
PubChem CID 75599
IUPAC Name 4-amino-2-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)N)C(=O)O

Specifications

Melting Point 145°C
Quantity 25 g
Formula Weight 167.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methoxybenzoic Acid
EINECSNumber (3)-1600&(9)-1469
TSCA No