accessibility menu, dialog, popup

UserName

CAS RN 2486-80-8

IUPAC Name: 4-amino-2-methoxybenzoic acid
Synonyms: 4-amino-2-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: OLJXRTRRJSMURJ-UHFFFAOYSA-N
SMILES: COC1=CC(N)=CC=C1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)