missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluorophenol 98.0+%, TCI America™

Catalog No. A21635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21635G 5 g
A21631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21635G Supplier TCI America Supplier No. A21635G

Chemical Identifiers

CAS 399-96-2
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00671760
InChI Key MXJQJURZHQZLNN-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxyaniline, 2-fluoro-4-aminophenol, phenol, 4-amino-2-fluoro, 4-amino-2-fluoro-phenol, zlchem 955, pubchem2831, acmc-1cswx, 4-amino-2-fluoro phenol
PubChem CID 2735918
IUPAC Name 4-amino-2-fluorophenol
SMILES C1=CC(=C(C=C1N)F)O

Specifications

Melting Point 164°C
Quantity 5 g
Formula Weight 127.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-fluorophenol