accessibility menu, dialog, popup

UserName

CAS RN 399-96-2

IUPAC Name: 4-amino-2-fluorophenol
Synonyms: 4-amino-2-fluorophenol
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: MXJQJURZHQZLNN-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results