missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluorobenzoic Acid 98.0+%, TCI America™

$171.49 - $680.38

Chemical Identifiers

CAS 446-31-1
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD01569397
InChI Key QHERSCUZBKDVOC-UHFFFAOYSA-N
Synonym 2-fluoro-4-aminobenzoic acid, 4-amino-2-fluorobenzoicacid, 4-amino-2-fluorobenzenecarboxylic acid, 4-amino-2-fluoro-benzoic acid, 4-carboxy-3-fluoroaniline, benzoic acid, 4-amino-2-fluoro, pubchem4693, acmc-209jz6, ksc237k0f, 4-amino-2-fluoro benzoic acid
PubChem CID 302680
IUPAC Name 4-amino-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1N)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A31645G
View Documents
TCI America
A31645G
5 g N/A
Only null left
 
A316425G
View Documents
TCI America
A316425G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

446-31-1
155.128
QHERSCUZBKDVOC-UHFFFAOYSA-N
302680
C1=CC(=C(C=C1N)F)C(=O)O
C7H6FNO2
MFCD01569397
2-fluoro-4-aminobenzoic acid, 4-amino-2-fluorobenzoicacid, 4-amino-2-fluorobenzenecarboxylic acid, 4-amino-2-fluoro-benzoic acid, 4-carboxy-3-fluoroaniline, benzoic acid, 4-amino-2-fluoro, pubchem4693, acmc-209jz6, ksc237k0f, 4-amino-2-fluoro benzoic acid
4-amino-2-fluorobenzoic acid

Specifications

446-31-1
White-Yellow
MFCD01569397
2-fluoro-4-aminobenzoic acid, 4-amino-2-fluorobenzoicacid, 4-amino-2-fluorobenzenecarboxylic acid, 4-amino-2-fluoro-benzoic acid, 4-carboxy-3-fluoroaniline, benzoic acid, 4-amino-2-fluoro, pubchem4693, acmc-209jz6, ksc237k0f, 4-amino-2-fluoro benzoic acid
C1=CC(=C(C=C1N)F)C(=O)O
155.128
155.13
Crystalline Powder
210°C
C7H6FNO2
5 g
QHERSCUZBKDVOC-UHFFFAOYSA-N
4-amino-2-fluorobenzoic acid
302680
≥98.0% (GC,T)
4-Amino-2-fluorobenzoic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'