accessibility menu, dialog, popup

UserName

CAS RN 446-31-1

IUPAC Name: 4-amino-2-fluorobenzoic acid
Synonyms: 2-fluoro-4-aminobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: QHERSCUZBKDVOC-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(O)=O)C(F)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)