missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. A31645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31645G 5 g
A316425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31645G Supplier TCI America Supplier No. A31645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446-31-1
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD01569397
InChI Key QHERSCUZBKDVOC-UHFFFAOYSA-N
Synonym 2-fluoro-4-aminobenzoic acid, 4-amino-2-fluorobenzoicacid, 4-amino-2-fluorobenzenecarboxylic acid, 4-amino-2-fluoro-benzoic acid, 4-carboxy-3-fluoroaniline, benzoic acid, 4-amino-2-fluoro, pubchem4693, acmc-209jz6, ksc237k0f, 4-amino-2-fluoro benzoic acid
PubChem CID 302680
IUPAC Name 4-amino-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1N)F)C(=O)O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-fluorobenzoic Acid