missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorophenol 98.0+%, TCI America™

Catalog No. p-7117474
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A14965G 5 g
A149625G 25 g
2 options

Catalog No. A14965G

Supplier: TCI America A14965G

Only null left

Chemical Identifiers

CAS 3964-52-1
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD00128891
InChI Key ZYZQSCWSPFLAFM-UHFFFAOYSA-N
Synonym 3-chloro-4-hydroxyaniline, 2-chloro-4-aminophenol, phenol, 4-amino-2-chloro, 4-amino-2-chloro-phenol, unii-2z6zt3b98u, o-chloro-p-aminophenol, pubchem18907, salzsaurem p-aminophenol, acmc-1aiqv, 3-chloro4-hydroxyaniline
PubChem CID 77578
IUPAC Name 4-amino-2-chlorophenol
SMILES NC1=CC=C(O)C(Cl)=C1

Specifications

Melting Point 153°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chlorophenol
EINECSNumber (3)-0906
TSCA No
Recommended Storage Refrigerator