accessibility menu, dialog, popup

UserName

CAS RN 3964-52-1

IUPAC Name: 4-amino-2-chlorophenol
Synonyms: 4-amino-2-chlorophenol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: ZYZQSCWSPFLAFM-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C(Cl)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)