missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzoic Acid 97.0+%, TCI America™

Catalog No. A09515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09515G 5 g
A095125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09515G Supplier TCI America Supplier No. A09515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2457-76-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007772
InChI Key MBDUKNCPOPMRJQ-UHFFFAOYSA-N
Synonym 2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
PubChem CID 17154
ChEBI CHEBI:59472
IUPAC Name 4-amino-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1N)Cl)C(=O)O

Specifications

Melting Point 211°C
Quantity 5 g
Formula Weight 171.58
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chlorobenzoic Acid
EINECSNumber (3)-1480
RTECSNumber DG1575000
TSCA Yes