accessibility menu, dialog, popup

UserName

CAS RN 2457-76-3

IUPAC Name: 4-amino-2-chlorobenzoic acid
Synonyms: 4-amino-2-chlorobenzoic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: MBDUKNCPOPMRJQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(O)=O)C(Cl)=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)