missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-bromophenol 98.0+%, TCI America™

Catalog No. A26471G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26471G Supplier TCI America Supplier No. A26471G
Only null left

Chemical Identifiers

CAS 16750-67-7
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD06656565
InChI Key CBQJZWGBFZAUEV-UHFFFAOYSA-N
Synonym 3-Bromo-4-hydroxyaniline
PubChem CID 14440265
IUPAC Name 4-amino-2-bromophenol
SMILES NC1=CC(Br)=C(O)C=C1

Specifications

Melting Point 163°C
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-bromophenol