accessibility menu, dialog, popup

UserName

CAS RN 16750-67-7

NH2BrOH

CAS RN 16750-67-7

IUPAC Name: 4-amino-2-bromophenol
Synonyms: 4-amino-2-bromophenol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: CBQJZWGBFZAUEV-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results