missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-diphenylphenol 98.0+%, TCI America™

Catalog No. A1269500MG
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1269500MG 500 mg
A12691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1269500MG Supplier TCI America Supplier No. A1269500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50432-01-4
Molecular Formula C18H15NO
Molecular Weight (g/mol) 261.324
MDL Number MFCD00034070
InChI Key YCOUFOVMXBWYIX-UHFFFAOYSA-N
Synonym 5′C-Amino-2′C-hydroxy-m-terphenyl
PubChem CID 621555
IUPAC Name 4-amino-2,6-diphenylphenol
SMILES C1=CC=C(C=C1)C2=CC(=CC(=C2O)C3=CC=CC=C3)N

Specifications

Melting Point 148°C
Color Red-White
Quantity 500 mg
Formula Weight 261.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-diphenylphenol