missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dichlorophenol 98.0+%, TCI America™

Catalog No. A087825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A087825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A087825G Supplier TCI America Supplier No. A087825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5930-28-9
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00007875
InChI Key KGEXISHTCZHGFT-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-aminophenol, phenol, 4-amino-2,6-dichloro, 2,6-dichloro-p-aminophenol, 3,5-dichloro-4-hydroxyaniline, unii-i0l4csx5n0, i0l4csx5n0, 2,6-dichloro-4-amino phenol, 4-amino-2,6-dichloro-phenol, acmc-209mc4, dsstox_cid_20720
PubChem CID 80037
IUPAC Name 4-amino-2,6-dichlorophenol
SMILES NC1=CC(Cl)=C(O)C(Cl)=C1

Specifications

Melting Point 167°C
Quantity 25 g
Formula Weight 178.01
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-dichlorophenol
EINECSNumber (3)-3818
RTECSNumber SJ5774500
TSCA Yes