accessibility menu, dialog, popup

UserName

CAS RN 5930-28-9

IUPAC Name: 4-amino-2,6-dichlorophenol
Synonyms: phenol, 4-amino-2,6-dichloro-
Molecular Weight (g/mol): 178.01
Molecular Formula: C6H5Cl2NO
InChi Key: KGEXISHTCZHGFT-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(O)C(Cl)=C1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)