missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,5-xylenol 97.0+%, TCI America™
$209.84 - $696.92
Chemical Identifiers
| CAS | 3096-71-7 |
|---|---|
| Molecular Formula | C8H11NO |
| Molecular Weight (g/mol) | 137.182 |
| InChI Key | JSWVCUXQICMATE-UHFFFAOYSA-N |
| Synonym | 4-amino-2,5-xylenol, phenol, 4-amino-2,5-dimethyl, unii-rf4r93709t, 2,5-dimethyl-4-aminophenol, 4-amino-2,5-dimethyl-phenol, 4-hydroxy-2,5-dimethylaniline, acmc-209hjo, 2,5-dimethy-4-aminophenol, 2,5-xylenol, 4-amino, ksc495q0r |
| PubChem CID | 76544 |
| IUPAC Name | 4-amino-2,5-dimethylphenol |
| SMILES | CC1=CC(=C(C=C1O)C)N |
Chemical Identifiers
| 3096-71-7 | |
| 137.182 | |
| 4-amino-2,5-xylenol, phenol, 4-amino-2,5-dimethyl, unii-rf4r93709t, 2,5-dimethyl-4-aminophenol, 4-amino-2,5-dimethyl-phenol, 4-hydroxy-2,5-dimethylaniline, acmc-209hjo, 2,5-dimethy-4-aminophenol, 2,5-xylenol, 4-amino, ksc495q0r | |
| 4-amino-2,5-dimethylphenol |
| C8H11NO | |
| JSWVCUXQICMATE-UHFFFAOYSA-N | |
| 76544 | |
| CC1=CC(=C(C=C1O)C)N |
Specifications
| 3096-71-7 | |
| C8H11NO | |
| 4-amino-2,5-xylenol, phenol, 4-amino-2,5-dimethyl, unii-rf4r93709t, 2,5-dimethyl-4-aminophenol, 4-amino-2,5-dimethyl-phenol, 4-hydroxy-2,5-dimethylaniline, acmc-209hjo, 2,5-dimethy-4-aminophenol, 2,5-xylenol, 4-amino, ksc495q0r | |
| CC1=CC(=C(C=C1O)C)N | |
| 137.182 | |
| 137.18 | |
| Crystalline Powder |
| 238°C | |
| 5 g | |
| JSWVCUXQICMATE-UHFFFAOYSA-N | |
| 4-amino-2,5-dimethylphenol | |
| 76544 | |
| ≥97.0% (GC,T) | |
| 4-Amino-2,5-xylenol |
Safety and Handling
TSCA : No