CAS RN 3096-71-7
IUPAC Name:
4-amino-2,5-dimethylphenol
Synonyms:
4-amino-2,5-dimethylphenol
Molecular Weight (g/mol):
137.18
Molecular Formula:
C8H11NO
InChi Key:
JSWVCUXQICMATE-UHFFFAOYSA-N
SMILES:
CC1=CC(O)=C(C)C=C1N
1
–
1
of
1
results
1
4-Amino-2,5-xylenol 97.0+%, TCI America™
CAS: 3096-71-7 Molecular Formula: C8H11NO Molecular Weight (g/mol): 137.182 InChI Key: JSWVCUXQICMATE-UHFFFAOYSA-N Synonym: 4-amino-2,5-xylenol,phenol, 4-amino-2,5-dimethyl,unii-rf4r93709t,2,5-dimethyl-4-aminophenol,4-amino-2,5-dimethyl-phenol,4-hydroxy-2,5-dimethylaniline,acmc-209hjo,2,5-dimethy-4-aminophenol,2,5-xylenol, 4-amino,ksc495q0r PubChem CID: 76544 IUPAC Name: 4-amino-2,5-dimethylphenol SMILES: CC1=CC(=C(C=C1O)C)N