missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,5-xylenol 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 238°C |
| Quantity | 5 g |
| Formula Weight | 137.18 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-2,5-xylenol |
Safety and Handling
| TSCA | No |