missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,5-difluorobenzonitrile 98.0+%, TCI America™
$55.92 - $240.46
Chemical Identifiers
| CAS | 112279-61-5 |
|---|---|
| Molecular Formula | C7H4F2N2 |
| Molecular Weight (g/mol) | 154.12 |
| MDL Number | MFCD00190102 |
| InChI Key | LAPGMTOHOQPDGI-UHFFFAOYSA-N |
| Synonym | 2,5-difluoro-4-aminobenzonitrile, 4-cyano-2,5-difluoroaniline, 4-amino-2,5-difluoro-benzonitrile, 4-amino-2,5-difluorobenzenecarbonitrile, 4-cyano, pubchem3536, ksc507o5b, 4-cyano-2,5-difluoro-aniline, timtec-bb sbb028311, buttpark 100\07-75 |
| PubChem CID | 2735900 |
| IUPAC Name | 4-amino-2,5-difluorobenzonitrile |
| SMILES | C1=C(C(=CC(=C1F)N)F)C#N |
Chemical Identifiers
| 112279-61-5 | |
| 154.12 | |
| LAPGMTOHOQPDGI-UHFFFAOYSA-N | |
| 2735900 | |
| C1=C(C(=CC(=C1F)N)F)C#N |
| C7H4F2N2 | |
| MFCD00190102 | |
| 2,5-difluoro-4-aminobenzonitrile, 4-cyano-2,5-difluoroaniline, 4-amino-2,5-difluoro-benzonitrile, 4-amino-2,5-difluorobenzenecarbonitrile, 4-cyano, pubchem3536, ksc507o5b, 4-cyano-2,5-difluoro-aniline, timtec-bb sbb028311, buttpark 100\07-75 | |
| 4-amino-2,5-difluorobenzonitrile |
Specifications
| 112279-61-5 | |
| Yellow | |
| MFCD00190102 | |
| 3439 | |
| LAPGMTOHOQPDGI-UHFFFAOYSA-N | |
| 4-amino-2,5-difluorobenzonitrile | |
| 2735900 | |
| ≥98.0% (GC) | |
| 4-Amino-2,5-difluorobenzonitrile |
| 97°C | |
| C7H4F2N2 | |
| 1 g | |
| 2,5-difluoro-4-aminobenzonitrile, 4-cyano-2,5-difluoroaniline, 4-amino-2,5-difluoro-benzonitrile, 4-amino-2,5-difluorobenzenecarbonitrile, 4-cyano, pubchem3536, ksc507o5b, 4-cyano-2,5-difluoro-aniline, timtec-bb sbb028311, buttpark 100\07-75 | |
| C1=C(C(=CC(=C1F)N)F)C#N | |
| 154.12 | |
| 154.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No