missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,5-difluorobenzonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 97°C |
| Color | Yellow |
| Quantity | 1 g |
| UN Number | 3439 |
| Formula Weight | 154.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-2,5-difluorobenzonitrile |
Safety and Handling
| TSCA | No |