missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,3,5,6-tetrafluorobenzamide 98.0+%, TCI America™

Catalog No. A09491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09491G 1 g
A09495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09491G Supplier TCI America Supplier No. A09491G

Chemical Identifiers

CAS 1548-74-9
Molecular Formula C7H4F4N2O
Molecular Weight (g/mol) 208.116
MDL Number MFCD00008000
InChI Key CAERPAFTLPIDJT-UHFFFAOYSA-N
PubChem CID 73769
IUPAC Name 4-amino-2,3,5,6-tetrafluorobenzamide
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C(=O)N

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 1 g
Synonym 4-aminotetrafluorobenzamide, acmc-1cujp, 4-amino-2,3,5,6-tetrafluorobenzamid, 2,3,5,6-tetrafluoro-4-aminobenzamide, benzamide,4-amino-2,3,5,6-tetrafluoro, benzamide, 4-amino-2,3,5,6-tetrafluoro
Formula Weight 208.12
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluorobenzamide