missing translation for 'onlineSavingsMsg'
Learn More

4-Allylpyrocatechol 98.0+%, TCI America™

Catalog No. p-7118163
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A294710MG 10 mg
A2947100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A294710MG Supplier TCI America Supplier No. A294710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1126-61-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD01548379
InChI Key FHEHIXJLCWUPCZ-UHFFFAOYSA-N
Synonym 4-Allyl-1,2-benzenediol, 4-Allyl-1,2-dihydroxybenzene, 4-Allylcatechol, Hydroxychavicol
PubChem CID 70775
IUPAC Name 4-prop-2-enylbenzene-1,2-diol
SMILES C=CCC1=CC(=C(C=C1)O)O

Specifications

Melting Point 44°C
Boiling Point 123°C
Quantity 10 mg
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Allylpyrocatechol
RTECSNumber CZ9029440
TSCA No
Recommended Storage Refrigerator