missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylpyridine 98.0+%, TCI America™

$87.60 - $529.40

Chemical Identifiers

CAS 1122-54-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006433
InChI Key WMQUKDQWMMOHSA-UHFFFAOYSA-N
Synonym 4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
PubChem CID 14282
IUPAC Name 1-pyridin-4-ylethanone
SMILES CC(=O)C1=CC=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A011325G
View Documents
TCI America
A011325G
25 g N/A
Only null left
 
A0113250G
View Documents
TCI America
A0113250G
250 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1122-54-9
121.139
WMQUKDQWMMOHSA-UHFFFAOYSA-N
14282
CC(=O)C1=CC=NC=C1
C7H7NO
MFCD00006433
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
1-pyridin-4-ylethanone

Specifications

1122-54-9
Yellow
C7H7NO
25 g
WMQUKDQWMMOHSA-UHFFFAOYSA-N
1-pyridin-4-ylethanone
14282
≥98.0% (GC,T)
4-Acetylpyridine
16°C
214°C
MFCD00006433
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
CC(=O)C1=CC=NC=C1
121.139
121.14
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3706

RTECSNumber : OB5426000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'