missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylpyridine 98.0+%, TCI America™

Supplier:  TCI America A011325G

 View more versions of this product

Catalog No. A011325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Acetylpyridine
16°C
214°C
MFCD00006433
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
CC(=O)C1=CC=NC=C1
121.139
121.14
Liquid
1122-54-9
Yellow
C7H7NO
25 g
WMQUKDQWMMOHSA-UHFFFAOYSA-N
1-pyridin-4-ylethanone
14282
≥98.0% (GC,T)

Chemical Identifiers

1122-54-9
121.139
WMQUKDQWMMOHSA-UHFFFAOYSA-N
14282
CC(=O)C1=CC=NC=C1
C7H7NO
MFCD00006433
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
1-pyridin-4-ylethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3706

RTECSNumber : OB5426000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions