missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetylbenzo-18-crown 6-Ether 97.0+%, TCI America™

Catalog No. A16041G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16041G Supplier TCI America Supplier No. A16041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41855-35-0
Molecular Formula C18H26O7
Molecular Weight (g/mol) 354.399
MDL Number MFCD00188456
InChI Key GOVDFSVHYMATAJ-UHFFFAOYSA-N
Synonym 2,3-(4-Acetylbenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene
PubChem CID 3660800
IUPAC Name 1-(2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-trien-20-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)OCCOCCOCCOCCOCCO2

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 1 g
Formula Weight 354.40
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Acetylbenzo-18-crown 6-Ether