accessibility menu, dialog, popup

UserName

CAS RN 41855-35-0

IUPAC Name: 1-(2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecin-18-yl)ethan-1-one
Synonyms: BENZO18CROWN6ethermacetyl
Molecular Weight (g/mol): 354.4
Molecular Formula: C18H26O7
InChi Key: GOVDFSVHYMATAJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=C(OCCOCCOCCOCCOCCO2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results