missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. A28181G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28181G 1 g
A2818200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28181G Supplier TCI America Supplier No. A28181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 177490-82-3
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD09027198
InChI Key VILXJXDXZGKJLU-UHFFFAOYSA-N
Synonym 4-acetoxyphenyl boronic acid, 4-acetoxyphenylboronic acid, 4-acetoxybenzeneboronic acid, 4-acetyloxy phenylboronic acid, boronic acid, 4-acetyloxy phenyl, boronic acid,b-4-acetyloxy phenyl, 4-boronophenyl acetate, pubchem20170, acmc-209ecy, 4-acetoxyphenyl boronicacid
PubChem CID 44119577
IUPAC Name [4-(acetyloxy)phenyl]boronic acid
SMILES CC(=O)OC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 1 g
Formula Weight 179.97
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetoxyphenylboronic Acid (contains varying amounts of Anhydride)