accessibility menu, dialog, popup

UserName

BOOH3CHOOH

CAS RN 177490-82-3

IUPAC Name: [4-(acetyloxy)phenyl]boronic acid
Synonyms: 4-(acetyloxy)phenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: VILXJXDXZGKJLU-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)