missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7035165
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52879MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH52879MD Supplier Thermo Scientific Chemicals Supplier No. H52879MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 177490-82-3
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD09027198
InChI Key VILXJXDXZGKJLU-UHFFFAOYSA-N
Synonym 4-acetoxyphenyl boronic acid, 4-acetoxyphenylboronic acid, 4-acetoxybenzeneboronic acid, 4-acetyloxy phenylboronic acid, boronic acid, 4-acetyloxy phenyl, boronic acid,b-4-acetyloxy phenyl, 4-boronophenyl acetate, pubchem20170, acmc-209ecy, 4-acetoxyphenyl boronicacid
PubChem CID 44119577
IUPAC Name (4-acetyloxyphenyl)boronic acid
SMILES CC(=O)OC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 220°C to 224°C
Quantity 250 mg
Formula Weight 179.97
Percent Purity 97%
Chemical Name or Material 4-Acetoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only