missing translation for 'onlineSavingsMsg'
Learn More

4'-Acetoxyacetanilide 99.0+%, TCI America™

Catalog No. A00675G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A00675G 5 g
A006725G 25 g
2 options

Catalog No. A00675G

Supplier: TCI America A00675G

Only null left

Chemical Identifiers

CAS 2623-33-8
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00059205
InChI Key UJAOSPFULOFZRR-UHFFFAOYSA-N
Synonym N,O-Diacetyl-4-aminophenol
PubChem CID 17499
IUPAC Name 4-acetamidophenyl acetate
SMILES CC(=O)NC1=CC=C(OC(C)=O)C=C1

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Acetoxyacetanilide