missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzyl alcohol, 97%

Catalog No. p-7032108
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3345403 1 g
AAH3345406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3345403 Supplier Thermo Scientific Chemicals Supplier No. H3345403

CAS: 16375-88-5 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16375-88-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00016868
InChI Key XEYORFKUJZEQCH-UHFFFAOYSA-N
Synonym 4-acetamidobenzyl alcohol, n-4-hydroxymethyl phenyl acetamide, 4-acetaminobenzyl alcohol, 4-acetamidobenzylalcohol, n-4-hydroxymethyl-phenyl-acetamide, p-acetaminobenzyl alcohol, acetamide, n-4-hydroxymethyl phenyl, 4-acetylamino benzyl alcohol
PubChem CID 152141
IUPAC Name N-[4-(hydroxymethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC=C(CO)C=C1

Specifications

Melting Point 118°C to 120°C
Quantity 1 g
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 4-Acetamidobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only