accessibility menu, dialog, popup

UserName

CAS RN 16375-88-5

IUPAC Name: N-[4-(hydroxymethyl)phenyl]acetamide
Synonyms: N-[4-(hydroxymethyl)phenyl]acetamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: XEYORFKUJZEQCH-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(CO)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)