missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-2-methylbenzoic Acid 96.0+%, TCI America™

Catalog No. p-7118100
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28385G 5 g
A283825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28385G Supplier TCI America Supplier No. A28385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103204-69-9
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD02258874
InChI Key AQPDTYYKDYMCTH-UHFFFAOYSA-N
Synonym 4-acetylamino-2-methylbenzoic acid, benzoic acid, 4-acetylamino-2-methyl, 4-acetamido-2-methyl-benzoic acid, acmc-20amqf, 4-acetamido-o-toluic acid, 4'-acetamido-2-methylbenzoicacid, 4-acetylamino-2-methyl-benzoic acid, o-toluic acid, 4-acetamido-6ci, n-acetyl-4-amino-2-methyl-benzoic acid
PubChem CID 2735224
IUPAC Name 4-acetamido-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)NC(=O)C)C(=O)O

Specifications

Melting Point 237°C
Color Yellow
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-2-methylbenzoic Acid