missing translation for 'onlineSavingsMsg'
Learn More

4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic Acid 98.0+%, TCI America™

Catalog No. p-7129976
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09251G 1 g
F09255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09251G Supplier TCI America Supplier No. F09251G
Only null left

Chemical Identifiers

CAS 164470-64-8
Molecular Formula C23H19NO4
Molecular Weight (g/mol) 373.41
MDL Number MFCD01074693
InChI Key JRHUROPSUJVMNH-UHFFFAOYSA-N
Synonym fmoc-4-amb-oh, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxycarbonylamino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid, 4-n-fmoc-aminomethyl-benzoic acid, 4-fmoc-aminomethyl benzoicacid, 4-fmoc-aminomethyl benzoic acid, fmoc-4-amb-oh fmoc-4-aminomethyl benzoic acid
PubChem CID 2756083
IUPAC Name 4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]benzoic acid
SMILES OC(=O)C1=CC=C(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1

Specifications

Melting Point 215°C
Color White
Quantity 1 g
Formula Weight 373.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic Acid
TSCA No
Recommended Storage Refrigerator