missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dihydroxy-2-methylpyrimidine 96.0+%, TCI America™
Supplier: TCI America D258925G
Specifications
| 4,6-Dihydroxy-2-methylpyrimidine | |
| C5H6N2O2 | |
| 25 g | |
| BPSGVKFIQZZFNH-UHFFFAOYSA-N | |
| 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one | |
| 222672 | |
| ≥96.0% (T) |
| 40497-30-1 | |
| MFCD00006104,MFCD00205627,MFCD20486784 | |
| 4,6-dihydroxy-2-methylpyrimidine, 2-methylpyrimidine-4,6-diol, 2-methyl-4,6-dihydroxypyrimidine, 2-methyl-4,6-pyrimidinediol, 2-methyl-1h,5h-pyrimidine-4,5-dione, 4 1h-pyrimidinone, 6-hydroxy-2-methyl, 2-methyl-4,6-dihydroxy pyrimidine, 6-hydroxy-2-methyl-3h-pyrimidin-4-one, 6-hydroxy-2-methylpyrimidin-4 3h-one, 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one | |
| CC1=NC(O)=CC(=O)N1 | |
| 126.12 | |
| 126.12 | |
| Crystalline Powder |
Chemical Identifiers
| 40497-30-1 | |
| 126.12 | |
| BPSGVKFIQZZFNH-UHFFFAOYSA-N | |
| 222672 | |
| CC1=NC(O)=CC(=O)N1 |
| C5H6N2O2 | |
| MFCD00006104,MFCD00205627,MFCD20486784 | |
| 4,6-dihydroxy-2-methylpyrimidine, 2-methylpyrimidine-4,6-diol, 2-methyl-4,6-dihydroxypyrimidine, 2-methyl-4,6-pyrimidinediol, 2-methyl-1h,5h-pyrimidine-4,5-dione, 4 1h-pyrimidinone, 6-hydroxy-2-methyl, 2-methyl-4,6-dihydroxy pyrimidine, 6-hydroxy-2-methyl-3h-pyrimidin-4-one, 6-hydroxy-2-methylpyrimidin-4 3h-one, 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one | |
| 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one |
Safety and Handling
TSCA : No