missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-2-methylpyrimidine 96.0+%, TCI America™

Catalog No. p-7127384
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25895G 5 g
D258925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25895G Supplier TCI America Supplier No. D25895G

Chemical Identifiers

CAS 40497-30-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006104,MFCD00205627,MFCD20486784
InChI Key BPSGVKFIQZZFNH-UHFFFAOYSA-N
Synonym 4,6-dihydroxy-2-methylpyrimidine, 2-methylpyrimidine-4,6-diol, 2-methyl-4,6-dihydroxypyrimidine, 2-methyl-4,6-pyrimidinediol, 2-methyl-1h,5h-pyrimidine-4,5-dione, 4 1h-pyrimidinone, 6-hydroxy-2-methyl, 2-methyl-4,6-dihydroxy pyrimidine, 6-hydroxy-2-methyl-3h-pyrimidin-4-one, 6-hydroxy-2-methylpyrimidin-4 3h-one, 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one
PubChem CID 222672
IUPAC Name 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one
SMILES CC1=NC(O)=CC(=O)N1

Specifications

Quantity 5 g
Formula Weight 126.12
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dihydroxy-2-methylpyrimidine