accessibility menu, dialog, popup

UserName

CH3NOHNOH

CAS RN 40497-30-1

IUPAC Name: 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one
Synonyms: 6-hydroxy-2-methyl-3H-pyrimidin-4-one
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: BPSGVKFIQZZFNH-UHFFFAOYSA-N
SMILES: CC1=NC(O)=CC(=O)N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)