missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-o-cresol 98.0+%, TCI America™

Catalog No. D03535G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D03535G Supplier TCI America Supplier No. D03535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1570-65-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00019988
InChI Key WJQZZLQMLJPKQH-UHFFFAOYSA-N
Synonym 4,6-dichloro-o-cresol, 4,6-dichloro-2-methylphenol, phenol, 2,4-dichloro-6-methyl, o-cresol, 4,6-dichloro, 4,6-dichlorocresol, phenol,2,4-dichloro-6-methyl, dichlorocresol, o-cresol,6-dichloro, 4,6-dichlor-o-kresol, acmc-1c0mk
PubChem CID 15292
IUPAC Name 2,4-dichloro-6-methylphenol
SMILES CC1=CC(=CC(=C1O)Cl)Cl

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 225°C
Quantity 5 g
Formula Weight 177.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-o-cresol