accessibility menu, dialog, popup

UserName

CAS RN 1570-65-6

IUPAC Name: 2,4-dichloro-6-methylphenol
Synonyms: 4,6-dichloro-O-cresol
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: WJQZZLQMLJPKQH-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(Cl)=C1O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)