missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 1780-26-3 |
|---|---|
| Molecular Formula | C5H4Cl2N2 |
| Molecular Weight (g/mol) | 163.01 |
| MDL Number | MFCD00090472 |
| InChI Key | FIMUTBLUWQGTIJ-UHFFFAOYSA-N |
| Synonym | 2-methyl-4,6-dichloropyrimidine, 4,6-dichloro-2-methyl-pyrimidine, 2-mdcp, pyrimidine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methylpyrimidiine, 4,6-dichlor-2-methylpyrimidin, 4,6-dichloro-2-methyl pyrimidine, pubchem7075, acmc-1bpg6, ksc490s3h |
| PubChem CID | 234997 |
| IUPAC Name | 4,6-dichloro-2-methylpyrimidine |
| SMILES | CC1=NC(=CC(=N1)Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC434260250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C5H4Cl2N2 | |
| MFCD00090472 | |
| 2-methyl-4,6-dichloropyrimidine, 4,6-dichloro-2-methyl-pyrimidine, 2-mdcp, pyrimidine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methylpyrimidiine, 4,6-dichlor-2-methylpyrimidin, 4,6-dichloro-2-methyl pyrimidine, pubchem7075, acmc-1bpg6, ksc490s3h | |
| 4,6-dichloro-2-methylpyrimidine |
Specifications
| 1780-26-3 | |
| 100.0 | |
| Authentic | |
| C5H4Cl2N2 | |
| 2-methyl-4,6-dichloropyrimidine, 4,6-dichloro-2-methyl-pyrimidine, 2-mdcp, pyrimidine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methylpyrimidiine, 4,6-dichlor-2-methylpyrimidin, 4,6-dichloro-2-methyl pyrimidine, pubchem7075, acmc-1bpg6, ksc490s3h | |
| CC1=NC(=CC(=N1)Cl)Cl | |
| 163.01 | |
| 163.01 | |
| 4, 6-Dichloro-2-methylpyrimidine |
| 96.0 | |
| 98°C | |
| 96% min. (GC) | |
| MFCD00090472 | |
| FIMUTBLUWQGTIJ-UHFFFAOYSA-N | |
| 4,6-dichloro-2-methylpyrimidine | |
| 234997 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning
RUO – Research Use Only