missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-methylpyrimidine, 97%, Thermo Scientific™

Catalog No. AC434260250
Change view
Click to view available options
Quantity:
25g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434260250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434260250 Supplier Thermo Scientific Chemicals Supplier No. 434260250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1780-26-3
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.01
MDL Number MFCD00090472
InChI Key FIMUTBLUWQGTIJ-UHFFFAOYSA-N
Synonym 2-methyl-4,6-dichloropyrimidine, 4,6-dichloro-2-methyl-pyrimidine, 2-mdcp, pyrimidine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methylpyrimidiine, 4,6-dichlor-2-methylpyrimidin, 4,6-dichloro-2-methyl pyrimidine, pubchem7075, acmc-1bpg6, ksc490s3h
PubChem CID 234997
IUPAC Name 4,6-dichloro-2-methylpyrimidine
SMILES CC1=NC(=CC(=N1)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25g
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 4, 6-Dichloro-2-methylpyrimidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

RUO – Research Use Only