accessibility menu, dialog, popup

UserName

CH3NClClN

CAS RN 1780-26-3

IUPAC Name: 4,6-dichloro-2-methylpyrimidine
Synonyms: 4,6-dichloro-2-methylpyrimidine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: FIMUTBLUWQGTIJ-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)